siRNA Details
| Gene Symbol: RUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135999.1 | NP_001129471.1 | 3, 4 | 3223 | ||
| NM_014806.3 | NP_055621.1 | 3, 4 | 2967 | |||
| NR_052015.2 | 3, 4 | 2642 | ||||
| XM_006716896.2 | XP_006716959.1 | 2, 3 | 2814 | |||
| GenBank mRNA | AB002373.2 | - | 3, 4 | 2809 | ||
| AK299805.1 | - | 3, 4 | 2913 | |||
| BC064843.1 | - | 2, 3 | 2011 | |||
| BC132766.1 | - | 2, 3 | 2752 | 40146788 | ||
| BC132770.1 | - | 2, 3 | 2752 | 40146792 | ||
| BC146654.1 | - | 3, 4 | 2941 | 40147826 | ||
| BC150211.1 | - | 3, 4 | 2554 | |||
| BC150262.1 | - | 3, 4 | 2820 | 9007205 | ||

RUSC2
Genomic Map