siRNA Details
| Gene Symbol: ZBTB39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014830.2 | NP_055645.1 | 2 | 895 | ||
| GenBank mRNA | AB002350.1 | - | 2 | 828 | ||
| AK292468.1 | - | 2 | 895 | |||
| BC140890.1 | - | 1 | 798 | 9021758 | ||
| BC146867.1 | - | 1 | 798 | 9056795 | ||
| BC152461.1 | - | 2 | 828 | 8862597 | ||

ZBTB39
Genomic Map