siRNA Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166252.1 | NP_001159724.1 | 6 | 1168 | ||
| NM_014839.4 | NP_055654.2 | 7 | 1342 | |||
| XM_011542498.2 | XP_011540800.1 | 7 | 1290 | |||
| GenBank mRNA | AB007924.3 | - | 7 | 1338 | ||
| AF357013.1 | - | 7 | 1115 | |||
| AF541281.1 | - | 7 | 1349 | |||
| AK291369.1 | - | 7 | 1142 | |||
| AK300361.1 | - | 6 | 912 | |||
| AL834390.1 | - | 6 | 855 | |||
| AY304518.1 | - | 7 | 1290 | |||
| BC042963.1 | - | 7 | 1160 | |||

PLPPR4
Genomic Map