siRNA Details
| Gene Symbol: TECPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172631.2 | NP_001166102.1 | 11 | 2869 | ||
| NM_014844.4 | NP_055659.2 | 11 | 2869 | |||
| GenBank mRNA | AB002295.1 | - | 8 | 2172 | ||
| AB002327.1 | - | 11 | 2818 | |||
| AK299690.1 | - | 4 | 269 | |||
| BC136647.1 | - | 10 | 2666 | 9020637 | ||
| BC142667.1 | - | 11 | 2702 | 40148157 | ||
| BC142715.1 | - | 11 | 2702 | 40148843 | ||
| BC151230.1 | - | 11 | 2842 | 8860116 | ||

TECPR2
Genomic Map