siRNA Details
| Gene Symbol: KIAA0196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014846.3 | NP_055661.3 | 10 | 1575 | ||
| XM_005251120.3 | XP_005251177.1 | 9 | 1265 | |||
| XM_011517409.1 | XP_011515711.1 | 10 | 1552 | |||
| XM_017014113.1 | XP_016869602.1 | 10 | 1780 | |||
| GenBank mRNA | AK223628.1 | - | 10 | 1523 | ||
| AK291032.1 | - | 10 | 1398 | |||
| AK296970.1 | - | 8 | 1083 | |||
| BC026951.1 | - | 9 | 1326 | 3882443 | ||
| BC106015.1 | - | 10 | 1464 | 6059577 | ||

KIAA0196
Genomic Map