siRNA Details
| Gene Symbol: MRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006039.4 | NP_006030.2 | 3 | 980 | ||
| XM_011525543.2 | XP_011523845.1 | 3 | 711 | |||
| GenBank mRNA | AB014609.2 | - | 3 | 980 | ||
| AF107292.1 | - | 3 | 621 | |||
| AF134838.1 | - | 3 | 702 | |||
| BC146647.1 | - | 3 | 699 | 40147137 | ||
| BC150212.1 | - | 3 | 684 | 40148946 | ||
| BC153884.1 | - | 3 | 980 | 8860500 | ||

MRC2
Genomic Map