siRNA Details
| Gene Symbol: RBM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146698.1 | NP_001140170.1 | 17 | 2258 | ||
| NM_001146699.1 | NP_001140171.1 | 17 | 2258 | |||
| NM_016196.3 | NP_057280.2 | 17 | 2258 | |||
| XM_017020280.1 | XP_016875769.1 | 16 | 2186 | |||
| XM_017020281.1 | XP_016875770.1 | 16 | 2186 | |||
| GenBank mRNA | AB014582.1 | - | 17 | 2193 | ||
| AK291444.1 | - | 17 | 2209 | |||
| AK314606.1 | - | 17 | 2258 | |||
| AL117547.1 | - | 17 | 2199 | |||
| BC004289.1 | - | 17 | 2197 | 3619715 | ||
| BC006137.1 | - | 17 | 2197 | 3677625 | ||

RBM19
Genomic Map