siRNA Details
| Gene Symbol: NCAPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014865.3 | NP_055680.3 | 7 | 668 | ||
| GenBank mRNA | AK022511.1 | - | 7 | 668 | ||
| AK125155.1 | - | 4 | 323 | |||
| AK128354.1 | - | 7 | 580 | |||
| AK303310.1 | - | 6 | 643 | |||
| BC016913.1 | - | 7 | 1400 | |||
| BC028182.1 | - | 7 | 680 | 5395775 | ||
| CN307514.1 | - | 6 | 646 | |||
| D63880.1 | - | 7 | 1421 | |||

NCAPD2
Genomic Map