siRNA Details
| Gene Symbol: PAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127460.2 | NP_001120932.1 | 9 | 1825 | ||
| NM_001166279.1 | NP_001159751.1 | 9 | 1754 | |||
| NM_014871.4 | NP_055686.3 | 9 | 1754 | |||
| GenBank mRNA | AB014610.2 | - | 9 | 1651 | ||
| AB107585.1 | - | 8 | 1448 | |||
| AK310822.1 | - | 9 | 1638 | |||
| BC024043.1 | - | 9 | 1667 | 3357890 | ||
| BC094885.1 | - | 9 | 1822 | 4823602 | ||
| BX648106.1 | - | 9 | 1742 | |||

PAN2
Genomic Map