siRNA Details
| Gene Symbol: HELZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014877.3 | NP_055692.2 | 32 | 5601 | ||
| XM_005257888.4 | XP_005257945.1 | 33 | 5730 | |||
| XM_005257889.4 | XP_005257946.1 | 32 | 5652 | |||
| XM_006722214.3 | XP_006722277.1 | 33 | 5733 | |||
| XM_006722215.3 | XP_006722278.1 | 31 | 5605 | |||
| XM_006722216.3 | XP_006722279.1 | 22 | 4435 | |||
| XM_011525544.2 | XP_011523846.1 | 33 | 5804 | |||
| XM_017025477.1 | XP_016880966.1 | 30 | 5521 | |||
| XM_017025478.1 | XP_016880967.1 | 33 | 5786 | |||
| GenBank mRNA | BC094881.1 | - | 30 | 5627 | 3076113 | |
| BC130582.1 | - | 32 | 5581 | 40146613 | ||
| BC144083.1 | - | 32 | 5584 | 9052603 | ||
| D29677.2 | - | 31 | 5559 | |||

HELZ
Genomic Map