siRNA Details
| Gene Symbol: FGF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005117.2 | NP_005108.1 | 3 | 1013 | ||
| GenBank mRNA | AB018122.1 | - | 3 | 550 | ||
| AF110400.1 | - | 3 | 1013 | |||
| AY358302.1 | - | 3 | 1013 | |||
| BC017664.1 | - | 3 | 681 | 3849343 | ||
| BT006729.1 | - | 3 | 550 | |||

FGF19
Genomic Map