siRNA Details
| Gene Symbol: TNFSF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005118.3 | NP_005109.2 | 2 | 347 | ||
| GenBank mRNA | AF520785.1 | - | 2 | 353 | ||
| AK291642.1 | - | 2 | 258 | |||
| AK313721.1 | - | 2 | 322 | |||
| BC069435.1 | - | 2 | 302 | 7262219 | ||
| BC074940.2 | - | 2 | 271 | 30915494 | ||
| BC074941.2 | - | 2 | 271 | 30915271 | ||
| BC104462.1 | - | 2 | 346 | 40030800 | ||
| BC104463.1 | - | 2 | 347 | 40030803 | ||

TNFSF15
Genomic Map