siRNA Details
| Gene Symbol: MED13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005121.2 | NP_005112.2 | 16 | 3660 | ||
| XM_011525551.2 | XP_011523853.1 | 16 | 4361 | |||
| XM_011525552.2 | XP_011523854.1 | 16 | 4361 | |||
| XM_011525553.2 | XP_011523855.1 | 13 | 3075 | |||
| GenBank mRNA | AB011165.2 | - | 13 | 3078 | ||
| AF117754.1 | - | 17 | 3660 | |||
| BC140891.1 | - | 16 | 3634 | 9021759 | ||

MED13
Genomic Map