siRNA Details
| Gene Symbol: CLEC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005127.2 | NP_005118.2 | 5 | 1061 | ||
| GenBank mRNA | AB015628.1 | - | 5 | 554 | ||
| AK313916.1 | - | 5 | 565 | |||
| BC005254.1 | - | 5 | 555 | 3996700 | ||
| BG540007.1 | - | 1 | 300 | |||
| CB160326.1 | - | 3 | 284 | |||
| X96719.1 | - | 5 | 536 | |||

CLEC2B
Genomic Map