siRNA Details
| Gene Symbol: DOPEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320714.1 | NP_001307643.1 | 32 | 6278 | ||
| NM_005128.3 | NP_005119.2 | 32 | 6169 | |||
| XM_017028509.1 | XP_016883998.1 | 29 | 5672 | |||
| GenBank mRNA | AB023150.2 | - | 29 | 5638 | ||
| AJ237839.2 | - | 32 | 6175 | |||
| AK026478.1 | - | 11 | 1105 | |||
| BC012791.2 | - | 14 | 1790 | |||

DOPEY2
Genomic Map