siRNA Details
| Gene Symbol: DGCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173533.1 | NP_001167004.1 | 5 | 805 | ||
| NM_001173534.1 | NP_001167005.1 | 5 | 796 | |||
| NM_001184781.1 | NP_001171710.1 | 6 | 919 | |||
| NM_005137.2 | NP_005128.1 | 6 | 928 | |||
| NR_033674.1 | 5 | 813 | ||||
| GenBank mRNA | AK291670.1 | - | 6 | 886 | ||
| AK295697.1 | - | 5 | 771 | |||
| AK296337.1 | - | 5 | 754 | |||
| AK303130.1 | - | 6 | 838 | |||
| AK304382.1 | - | 5 | 763 | |||
| BC020339.1 | - | 6 | 875 | |||
| BC032430.1 | - | 6 | 864 | 5243412 | ||
| BC035425.1 | - | 6 | 875 | |||
| BC040500.1 | - | 6 | 923 | 5272271 | ||
| CR456433.1 | - | 6 | 700 | |||
| CR936871.1 | - | 7 | 1263 | |||
| D79985.1 | - | 6 | 865 | |||
| FJ032369.1 | - | 6 | 680 | |||
| L77560.1 | - | 6 | 829 | |||
| X83545.1 | - | 6 | 829 | |||
| X84076.1 | - | 6 | 826 | |||

DGCR2
Genomic Map