siRNA Details
| Gene Symbol: SCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169109.1 | NP_001162580.1 | 2 | 915 | ||
| NM_001169110.1 | NP_001162581.1 | 2 | 866 | |||
| NM_001169111.1 | NP_001162582.1 | 2 | 884 | |||
| NM_005138.2 | NP_005129.2 | 2 | 855 | |||
| GenBank mRNA | AL021683.1 | - | 2 | 793 | ||
| BC102024.1 | - | 2 | 783 | 40030584 | ||
| BC102025.1 | - | 2 | 783 | 40030589 | ||
| CR456569.1 | - | 1 | 713 | |||
| Gene Symbol: NCAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185011.1 | NP_001171940.1 | 20 | 2268 | ||
| NM_152299.3 | NP_689512.2 | 20 | 2265 | |||

SCO2
Genomic Map