siRNA Details
| Gene Symbol: TSSC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297658.1 | NP_001284587.1 | 4 | 1295 | ||
| NM_001297659.1 | NP_001284588.1 | 3 | 1143 | |||
| NM_001297660.1 | NP_001284589.1 | 4 | 1127 | |||
| NM_001297661.1 | NP_001284590.1 | 4 | 794 | |||
| NM_005706.3 | NP_005697.2 | 3 | 986 | |||
| XM_006718118.2 | XP_006718181.1 | 3 | 2025 | |||
| XM_011519830.2 | XP_011518132.1 | 4 | 1237 | |||
| GenBank mRNA | AF125568.1 | - | 2 | 676 | ||
| BC006091.1 | - | 3 | 699 | 3677524 | ||
| BC050616.1 | - | 4 | 794 | 5744058 | ||
| BX384913.2 | - | 4 | 855 | |||

TSSC4
Genomic Map