siRNA Details
| Gene Symbol: TSPAN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005723.3 | NP_005714.2 | 4 | 834 | ||
| XM_005262680.1 | XP_005262737.1 | 3 | 687 | |||
| GenBank mRNA | AF065389.1 | - | 4 | 746 | ||
| AK126702.1 | - | 1 | 3473 | |||
| AK295385.1 | - | 4 | 428 | |||
| AK315723.1 | - | 4 | 801 | |||
| BC009704.1 | - | 4 | 732 | 3895933 | ||
| CR541809.1 | - | 4 | 399 | |||
| CR541829.1 | - | 4 | 399 | |||

TSPAN5
Genomic Map