siRNA Details
Gene Symbol: KIF20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005733.2 | NP_005724.1 | 7 | 1219 | ||
GenBank mRNA | AF070672.1 | - | 6 | 750 | ||
AF153329.1 | - | 7 | 757 | |||
AK025790.1 | - | 7 | 778 | |||
AK296879.1 | - | 8 | 733 | |||
BC012999.2 | - | 7 | 757 | 3604291 |
