siRNA Details
| Gene Symbol: ZNF263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005741.4 | NP_005732.2 | 6 | 1955 | ||
| XM_006720831.1 | XP_006720894.1 | 6 | 2023 | |||
| XM_006720832.1 | XP_006720895.1 | 5 | 1945 | |||
| XM_006720833.1 | XP_006720896.1 | 4 | 1807 | |||
| XM_006720834.3 | XP_006720897.1 | 5 | 2078 | |||
| XM_011522344.1 | XP_011520646.1 | 6 | 2062 | |||
| XM_017022850.1 | XP_016878339.1 | 5 | 1501 | |||
| GenBank mRNA | AK295350.1 | - | 2 | 821 | ||
| AK312421.1 | - | 6 | 1937 | |||
| BC008805.2 | - | 6 | 1953 | 3945548 | ||
| D88827.1 | - | 6 | 1915 | |||

ZNF263
Genomic Map