siRNA Details
| Gene Symbol: CLEC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244755.1 | NP_001231684.1 | 2 | 421 | ||
| NM_005752.4 | NP_005743.4 | 3 | 642 | |||
| GenBank mRNA | AA399390.1 | - | 2 | 422 | ||
| AK313316.1 | - | 3 | 642 | |||
| AY358376.1 | - | 3 | 594 | |||
| BC104414.2 | - | 3 | 631 | 40003144 | ||
| BC104415.2 | - | 3 | 631 | 40003149 | ||

CLEC3A
Genomic Map