siRNA Details
| Gene Symbol: G3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005754.2 | NP_005745.1 | 11 | 1288 | ||
| NM_198395.1 | NP_938405.1 | 11 | 1257 | |||
| XM_006714749.3 | XP_006714812.1 | 11 | 1277 | |||
| XM_006714750.3 | XP_006714813.1 | 11 | 1216 | |||
| XM_017008922.1 | XP_016864411.1 | 11 | 1563 | |||
| XM_017008923.1 | XP_016864412.1 | 11 | 1532 | |||
| GenBank mRNA | AK130003.1 | - | 8 | 772 | ||
| AK222606.1 | - | 11 | 1249 | |||
| AK300098.1 | - | 10 | 1158 | |||
| AK303580.1 | - | 11 | 1345 | |||
| AK312572.1 | - | 11 | 1251 | |||
| BC006997.1 | - | 11 | 1251 | 3950738 | ||
| BC108278.1 | - | 11 | 1196 | 6051310 | ||
| BT019384.1 | - | 10 | 1117 | |||
| BX647869.1 | - | 10 | 1159 | |||
| BX648012.1 | - | 11 | 1263 | |||
| CR536535.1 | - | 10 | 1117 | |||
| U32519.1 | - | 11 | 1238 | |||

G3BP1
Genomic Map