siRNA Details
| Gene Symbol: CHST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166395.1 | NP_001159867.1 | 2 | 1325 | ||
| NM_005769.2 | NP_005760.1 | 2 | 1273 | |||
| GenBank mRNA | AF131235.1 | - | 2 | 1243 | ||
| AF149783.1 | - | 1 | 1154 | |||
| AF280088.1 | - | 2 | 1183 | |||
| AK026635.1 | - | 2 | 1133 | |||
| AK074746.1 | - | 2 | 1130 | |||
| BC035282.1 | - | 2 | 1139 | 5179826 | ||

CHST4
Genomic Map