siRNA Details
| Gene Symbol: RBM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005778.3 | NP_005769.1 | 4 | 393 | ||
| NR_036627.2 | 4 | 393 | ||||
| XM_006712917.1 | XP_006712980.1 | 4 | 393 | |||
| XM_011533261.1 | XP_011531563.1 | 4 | 393 | |||
| XM_011533262.1 | XP_011531564.1 | 4 | 393 | |||
| XM_017005503.1 | XP_016860992.1 | 4 | 375 | |||
| XM_017005504.1 | XP_016860993.1 | 4 | 393 | |||
| XM_017005505.1 | XP_016860994.1 | 4 | 347 | |||
| XM_017005506.1 | XP_016860995.1 | 3 | 271 | |||
| XM_017005507.1 | XP_016860996.1 | 3 | 271 | |||
| GenBank mRNA | AB208813.1 | - | 4 | 338 | ||
| AB586690.1 | - | 4 | 324 | |||
| AF091263.1 | - | 4 | 366 | |||
| AF103802.4 | - | 4 | 344 | |||
| AF107493.1 | - | 4 | 342 | |||
| AK297249.1 | - | 4 | 347 | |||
| AK314032.1 | - | 4 | 348 | |||
| AL701303.1 | - | 4 | 383 | |||
| BC002957.1 | - | 4 | 339 | |||
| DB080737.1 | - | 4 | 393 | |||
| KU178505.1 | - | 3 | 218 | |||
| KU178506.1 | - | 3 | 218 | |||
| U23946.1 | - | 4 | 287 | |||

RBM5
Genomic Map