siRNA Details
| Gene Symbol: CTDSPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008392.1 | NP_001008393.1 | 4, 5 | 383 | ||
| NM_005808.2 | NP_005799.2 | 3, 4 | 350 | |||
| XM_017005519.1 | XP_016861008.1 | 4, 5 | 409 | |||
| XM_017005520.1 | XP_016861009.1 | 4, 5 | 1727 | |||
| GenBank mRNA | AI143812.1 | - | 4, 5 | 383 | ||
| AI192993.1 | - | 3, 4 | 350 | |||
| AJ575644.1 | - | 3, 4 | 350 | |||
| AJ575645.1 | - | 4, 5 | 383 | |||
| AY279532.1 | - | 3, 4 | 328 | |||
| AY364238.1 | - | 3, 4 | 328 | |||
| D88153.1 | - | 4, 5 | 700 | |||

CTDSPL
Genomic Map