siRNA Details
| Gene Symbol: ANGPTL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021146.3 | NP_066969.1 | 4 | 1077 | ||
| GenBank mRNA | AB593026.1 | - | 4 | 1011 | ||
| AB593027.1 | - | 4 | 1078 | |||
| AK313716.1 | - | 4 | 1043 | |||
| AY358301.1 | - | 4 | 1000 | |||
| BC001881.1 | - | 4 | 1025 | 3544149 | ||
| BT009802.1 | - | 4 | 786 | |||
| Y16132.1 | - | 4 | 793 | |||

ANGPTL7
Genomic Map