siRNA Details
| Gene Symbol: ANGPTL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021146.3 | NP_066969.1 | 2, 3 | 767 | ||
| XM_017000004.1 | XP_016855493.1 | 2, 3 | 732 | |||
| GenBank mRNA | AB593026.1 | - | 2, 3 | 701 | ||
| AB593027.1 | - | 2, 3 | 768 | |||
| AK313716.1 | - | 2, 3 | 733 | |||
| AY358301.1 | - | 2, 3 | 690 | |||
| BC001881.1 | - | 2, 3 | 715 | 3544149 | ||
| BT009802.1 | - | 2, 3 | 476 | |||
| Y16132.1 | - | 2, 3 | 483 | |||

ANGPTL7
Genomic Map