siRNA Details
| Gene Symbol: CD96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005816.4 | NP_005807.1 | 13 | 1727 | ||
| NM_198196.2 | NP_937839.1 | 14 | 1775 | |||
| NR_134917.1 | 13 | 1724 | ||||
| XM_006713469.3 | XP_006713532.1 | 14 | 1733 | |||
| XM_006713470.3 | XP_006713533.1 | 13 | 1685 | |||
| XM_017005521.1 | XP_016861010.1 | 13 | 1547 | |||
| XM_017005522.1 | XP_016861011.1 | 11 | 1424 | |||
| GenBank mRNA | AK290719.1 | - | 13 | 1610 | ||
| AK315237.1 | - | 13 | 1609 | |||
| AL833681.1 | - | 14 | 1622 | |||

CD96
Genomic Map