siRNA Details
| Gene Symbol: SPRY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304990.1 | NP_001291919.1 | 3 | 693 | ||
| NM_005840.2 | NP_005831.1 | 2 | 837 | |||
| GenBank mRNA | AF041038.1 | - | 1 | 56 | ||
| AK289543.1 | - | 2 | 830 | |||
| BC069609.1 | - | 1 | 399 | 7262284 | ||
| BC074972.2 | - | 1 | 375 | 30915293 | ||
| BC074973.2 | - | 1 | 375 | 30915511 | ||

SPRY3
Genomic Map