siRNA Details
| Gene Symbol: SPRY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258038.1 | NP_001244967.1 | 3 | 539 | ||
| NM_001258039.1 | NP_001244968.1 | 2 | 283 | |||
| NM_005841.2 | NP_005832.1 | 2 | 460 | |||
| NM_199327.2 | NP_955359.1 | 2 | 293 | |||
| XM_005262686.1 | XP_005262743.1 | 3 | 1200 | |||
| XM_017007635.1 | XP_016863124.1 | 2 | 273 | |||
| GenBank mRNA | AK026960.1 | - | 2 | 440 | ||
| AL602431.1 | - | 3 | 539 | |||
| AV692593.1 | - | 2 | 279 | |||
| AY192146.1 | - | 2 | 288 | |||
| AY590694.1 | - | 1 | 91 | |||
| BC063856.1 | - | 2 | 422 | 6181674 | ||
| CN302060.1 | - | 2 | 263 | |||
| DA765300.1 | - | 3 | 510 | |||
| DB250100.1 | - | 2 | 460 | |||

SPRY1
Genomic Map