siRNA Details
| Gene Symbol: NET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047160.2 | NP_001040625.1 | 10 | 1224 | ||
| NM_005863.4 | NP_005854.2 | 8 | 1135 | |||
| NR_073040.1 | 8 | 1168 | ||||
| GenBank mRNA | AB209852.1 | - | 10 | 1170 | ||
| AJ010046.1 | - | 8 | 1068 | |||
| AK024919.1 | - | 8 | 1075 | |||
| AK294244.1 | - | 4 | 601 | |||
| AK299335.1 | - | 10 | 1180 | |||
| AK302687.1 | - | 10 | 1006 | |||
| AK304028.1 | - | 8 | 1110 | |||
| AK315566.1 | - | 8 | 1077 | |||
| BC010285.1 | - | 8 | 1066 | 3048980 | ||
| BC053553.1 | - | 10 | 1176 | 6137683 | ||
| BX537509.1 | - | 4 | 3773 | |||
| U02081.1 | - | 6 | 656 | |||

NET1
Genomic Map