siRNA Details
| Gene Symbol: NET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047160.2 | NP_001040625.1 | 10, 11 | 1328 | ||
| NM_005863.4 | NP_005854.2 | 8, 9 | 1239 | |||
| NR_073040.1 | 8, 9 | 1272 | ||||
| GenBank mRNA | AB209852.1 | - | 10, 11 | 1274 | ||
| AJ010046.1 | - | 8, 9 | 1172 | |||
| AK024919.1 | - | 8, 9 | 1179 | |||
| AK294244.1 | - | 4, 5 | 705 | |||
| AK299335.1 | - | 10, 11 | 1284 | |||
| AK302687.1 | - | 10, 11 | 1110 | |||
| AK304028.1 | - | 8, 9 | 1214 | |||
| AK315566.1 | - | 8, 9 | 1181 | |||
| BC010285.1 | - | 8, 9 | 1170 | 3048980 | ||
| BC053553.1 | - | 10, 11 | 1280 | 6137683 | ||
| BX537509.1 | - | 4, 5 | 3877 | |||
| U02081.1 | - | 6, 7 | 760 | |||

NET1
Genomic Map