siRNA Details
| Gene Symbol: EIF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005875.2 | NP_005866.1 | 2 | 359 | ||
| GenBank mRNA | AF064607.1 | - | 2 | 169 | ||
| AF077052.1 | - | 2 | 340 | |||
| AF263452.1 | - | 2 | 332 | |||
| AK223448.1 | - | 2 | 354 | |||
| AK311922.1 | - | 2 | 264 | |||
| BC006996.1 | - | 2 | 343 | 4044439 | ||
| BF667107.1 | - | 2 | 356 | |||
| CA335728.1 | - | 2 | 372 | |||
| CR542223.1 | - | 2 | 99 | |||

EIF1B
Genomic Map