siRNA Details
| Gene Symbol: LANCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136574.1 | NP_001130046.1 | 6 | 665 | ||
| NM_001136575.1 | NP_001130047.1 | 6 | 660 | |||
| NM_006055.2 | NP_006046.1 | 6 | 754 | |||
| XM_005246243.2 | XP_005246300.1 | 6 | 775 | |||
| GenBank mRNA | AK312381.1 | - | 6 | 643 | ||
| BC028685.2 | - | 6 | 665 | 4829574 | ||
| CR541777.1 | - | 5 | 590 | |||
| Y11395.1 | - | 6 | 694 | |||

LANCL1
Genomic Map