siRNA Details
| Gene Symbol: LANCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136574.1 | NP_001130046.1 | 7 | 868 | ||
| NM_001136575.1 | NP_001130047.1 | 7 | 863 | |||
| NM_006055.2 | NP_006046.1 | 7 | 957 | |||
| XM_005246243.2 | XP_005246300.1 | 7 | 978 | |||
| GenBank mRNA | AK312381.1 | - | 7 | 846 | ||
| BC028685.2 | - | 7 | 868 | 4829574 | ||
| CR541777.1 | - | 6 | 793 | |||
| Y11395.1 | - | 7 | 897 | |||

LANCL1
Genomic Map