siRNA Details
| Gene Symbol: CRISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190986.1 | NP_001177915.1 | 5 | 463 | ||
| NM_006061.2 | NP_006052.2 | 5 | 451 | |||
| GenBank mRNA | AK223380.1 | - | 5 | 355 | ||
| AK313387.1 | - | 5 | 384 | |||
| BC069580.1 | - | 4 | 337 | 7262396 | ||
| BC069602.1 | - | 4 | 339 | 7262408 | ||
| X95240.1 | - | 4 | 339 | |||

CRISP3
Genomic Map