siRNA Details
Gene Symbol: RRAGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006064.4 | NP_006055.3 | 6 | 1187 | ||
NM_016656.3 | NP_057740.2 | 7 | 1271 | |||
XM_005261973.4 | XP_005262030.1 | 7 | 608 | |||
XM_017029217.1 | XP_016884706.1 | 6 | 517 | |||
XM_017029218.1 | XP_016884707.1 | 7 | 1227 | |||
GenBank mRNA | AK289407.1 | - | 6 | 1120 | ||
AL831926.1 | - | 6 | 1126 | |||
BC034726.1 | - | 6 | 1118 | 3919252 | ||
X90530.1 | - | 7 | 1062 |
