siRNA Details
| Gene Symbol: CNPY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014255.5 | NP_055070.1 | 6 | 1075 | ||
| GenBank mRNA | AB015631.1 | - | 6 | 670 | ||
| AK312921.1 | - | 6 | 772 | |||
| AY032624.1 | - | 6 | 944 | |||
| AY359102.1 | - | 6 | 1068 | |||
| BC065015.1 | - | 6 | 710 | 6169229 | ||

CNPY2
Genomic Map