siRNA Details
| Gene Symbol: CNPY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190991.1 | NP_001177920.1 | 3 | 652 | ||
| NM_014255.5 | NP_055070.1 | 3 | 652 | |||
| GenBank mRNA | AB015631.1 | - | 3 | 247 | ||
| AF186113.1 | - | 3 | 206 | |||
| AK312921.1 | - | 3 | 349 | |||
| AY032624.1 | - | 3 | 521 | |||
| AY359102.1 | - | 3 | 645 | |||
| BC001027.2 | - | 3 | 343 | |||
| BC065015.1 | - | 3 | 287 | 6169229 | ||

CNPY2
Genomic Map