siRNA Details
| Gene Symbol: ABCA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080282.3 | NP_525021.3 | 22 | 3409 | ||
| GenBank mRNA | AL832004.2 | - | 22 | 3115 | ||
| AY028900.1 | - | 22 | 3409 | |||
| AY247065.1 | - | 22 | 3440 | |||
| BC136460.1 | - | 19 | 2614 | |||
| BC144124.1 | - | 19 | 2683 | |||

ABCA10
Genomic Map