siRNA Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016172.2 | NP_057256.2 | 8 | 1130 | ||
| GenBank mRNA | AF176796.1 | - | 8 | 1100 | ||
| BC004967.1 | - | 8 | 1130 | 3535682 | ||
| BC011822.1 | - | 8 | 956 | 3688526 | ||
| BX647463.1 | - | 1 | 5183 | |||

UBAC1
Genomic Map