siRNA Details
| Gene Symbol: MAD2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127325.1 | NP_001120797.1 | 5 | 507 | ||
| NM_006341.3 | NP_006332.3 | 5 | 474 | |||
| XM_011540507.1 | XP_011538809.1 | 5 | 335 | |||
| GenBank mRNA | AF072933.1 | - | 5 | 318 | ||
| AF139365.1 | - | 5 | 397 | |||
| AF157482.1 | - | 5 | 474 | |||
| AK027327.1 | - | 5 | 387 | |||
| AK094316.1 | - | 7 | 1216 | |||
| BC015244.1 | - | 4 | 288 | 3916742 | ||
| BI517654.1 | - | 5 | 509 | |||

MAD2L2
Genomic Map