siRNA Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278196.1 | NP_001265125.1 | 2 | 1515 | ||
| NM_006352.4 | NP_006343.2 | 1 | 2043 | |||
| NM_205768.2 | NP_991331.1 | 2 | 1646 | |||
| XM_005273006.2 | XP_005273063.1 | 3 | 1576 | |||
| XM_017000060.1 | XP_016855549.1 | 3 | 3676 | |||
| GenBank mRNA | AJ001388.1 | - | 1 | 1969 | ||
| AK291408.1 | - | 2 | 1623 | |||
| AK293892.1 | - | 3 | 1590 | |||
| BC036677.2 | - | 2 | 1622 | 5259205 | ||
| BC078175.1 | - | 1 | 2043 | |||
| U38896.1 | - | 2 | 1338 | |||
| X95072.1 | - | 1 | 1762 | |||

ZBTB18
Genomic Map