siRNA Details
| Gene Symbol: TM9SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014842.2 | NP_001014842.1 | 2 | 538 | ||
| NM_006405.6 | NP_006396.2 | 2 | 538 | |||
| GenBank mRNA | AK292110.1 | - | 2 | 227 | ||
| BC010856.2 | - | 1 | 147 | 3899246 | ||
| BM922952.1 | - | 489 | ||||
| BX161390.1 | - | 2 | 458 | |||
| BX161494.1 | - | 2 | 420 | |||
| U94831.1 | - | 1 | 145 | |||

TM9SF1
Genomic Map