siRNA Details
| Gene Symbol: TM9SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014842.2 | NP_001014842.1 | 3 | 900 | ||
| NM_001289006.1 | NP_001275935.1 | 3 | 694 | |||
| NM_006405.6 | NP_006396.2 | 3 | 900 | |||
| GenBank mRNA | AK292110.1 | - | 3 | 589 | ||
| AK304642.1 | - | 3 | 595 | |||
| BC010856.2 | - | 2 | 509 | 3899246 | ||
| BX161390.1 | - | 3 | 820 | |||
| BX161494.1 | - | 3 | 782 | |||
| U94831.1 | - | 2 | 507 | |||

TM9SF1
Genomic Map