siRNA Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006417.4 | NP_006408.3 | 7 | 1203 | ||
| XM_011540516.2 | XP_011538818.1 | 7 | 2441 | |||
| XM_017000083.1 | XP_016855572.1 | 7 | 2441 | |||
| GenBank mRNA | AK302527.1 | - | 7 | 1164 | ||
| AK316278.1 | - | 6 | 696 | |||
| AK316476.1 | - | 7 | 1161 | |||
| BC022870.1 | - | 7 | 1168 | 4071404 | ||
| CA426666.1 | - | 3 | 536 | |||

IFI44
Genomic Map