siRNA Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006425.4 | NP_006416.3 | 11 | 1161 | ||
| XM_005265793.3 | XP_005265850.1 | 11 | 1508 | |||
| XM_011534418.2 | XP_011532720.1 | 12 | 1211 | |||
| GenBank mRNA | AF101074.1 | - | 10 | 1006 | ||
| AK024875.1 | - | 11 | 1081 | |||
| AK314564.1 | - | 11 | 1081 | |||
| BC010634.1 | - | 11 | 1087 | 3871882 | ||

SLU7
Genomic Map