siRNA Details
| Gene Symbol: DPYSL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006426.2 | NP_006417.2 | 10 | 1245 | ||
| XM_005252658.3 | XP_005252715.1 | 10 | 1234 | |||
| XM_011539125.2 | XP_011537427.1 | 11 | 1248 | |||
| XM_017015482.1 | XP_016870971.1 | 10 | 2509 | |||
| XM_017015483.1 | XP_016870972.1 | 11 | 2662 | |||
| GenBank mRNA | AB006713.1 | - | 10 | 1239 | ||
| BC029280.1 | - | 10 | 1129 | |||
| BC136329.1 | - | 10 | 1114 | 9020316 | ||
| BC136330.1 | - | 10 | 1114 | 9020317 | ||
| Y10976.1 | - | 10 | 1122 | |||

DPYSL4
Genomic Map