siRNA Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006470.3 | NP_006461.3 | 9 | 2121 | ||
| GenBank mRNA | AK002035.1 | - | 6 | 1587 | ||
| AK056026.1 | - | 7 | 2402 | |||
| AK314557.1 | - | 6 | 1601 | |||
| BC001564.1 | - | 6 | 1451 | |||
| BC015674.2 | - | 5 | 1140 | |||
| BC053514.1 | - | 9 | 2121 | 6263729 | ||
| BC067096.1 | - | 9 | 2121 | 6088764 | ||
| CR457136.1 | - | 6 | 1564 | |||

TRIM16
Genomic Map